Value Plastics Female Luer Thread Union Fittings
Luer fittings allow for a simple yet secure connection
  • Quick and easy assembly and disassembly without the need for tools
  • Thread design provides a secure closure—preventing contamination
  • Conforms to ISO-594-1 and ISO 594-2 standards
The Female Luer Thread Union Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The thread feature is built into the female luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. Adapters are available in various materials to accommodate various applications and compliance requirements.
3,200.00 - ₹3,400.00INR / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Regulatory Approvals Description
Availability Pricing
Value Plastics Luer Fitting, Straight Union, Polypropylene, Female Luer Thread; 25/Pk 5012057 Mfr # FTLLC-6005
COMPARE
Polypropylene Non-DEHP Luer Fitting, Straight Union, Polypropylene, Female Luer Thread; 25/Pk
₹3,400.00INR / Pkg of 25
Value Plastics Luer Fitting, Straight Union, Clear Polycarbonate, Female Luer Thread ; 25/Pk 5012066 Mfr # FTLLC-9
COMPARE
Polycarbonate Non-Phthalate Luer Fitting, Straight Union, Clear Polycarbonate, Female Luer Thread ; 25/Pk
₹3,200.00INR / Pkg of 25