Vaisala HUMICAP® Relative Humidity and Temperature Transmitters
Designed for industrial and HVAC applications
  • Easily and accurately measure humidity and temperature
Vaisala HUMICAP® humidity and temperature sensors have excellent accuracy and long-term stability, negligible hysteresis, and resistance to dust and most chemicals. Two-wire transmitters monitor your humidity or RH/temperature conditions with ±2% RH accuracy. All wall-mount models have ABS plastic housing; duct-mount models have aluminum housing.
123,000.00 - ₹162,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RH Range (%) Max Temperature (° F) Mounting Type
Availability Pricing
Vaisala HMW92 Relative Humidity/Temperature Transmitter, Wall-Mount WW-03334-25 Mfr # HMW92
COMPARE
100 131 Wall
₹123,000.00INR / Each
Vaisala HMD62 Relative Humidity/Temperature Transmitter, Duct-Mount WW-03334-35 Mfr # HMD62
COMPARE
100 176 Duct
₹162,000.00INR / Each
Vaisala HMW92D Relative Humidity/Temperature Transmitter, Wall-Mount with Display WW-03334-55 Mfr # HMW92D
COMPARE
100 131 Wall
₹123,000.00INR / Each