United Scientific Supplies Filtering Glassware
Heavy-duty filtering flasks optimize your vacuum filtration process
  • Easy-to-read white enamel graduations
  • Heavy-wall borosilicate glass with sturdy rim for enhanced durability
  • Autoclavable
These heavy-wall flasks with tubulation are constructed of durable borosilicate glass designed to give maximum mechanical strength. For convenience, the flasks are graduated to show approximate capacity. The neck finish and heavy-duty rim affords an excellent fit for rubber stoppers.
2,800.00 - ₹63,100.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Height (in) Description
Availability Pricing
United Scientific Supplies Filtering Flask, Borosilicate Glass, 50 mL; Pk/6 WW-34509-45 Mfr # FG5340-50
COMPARE
50 3 11/32 Filtering Flask, Borosilicate Glass, 50 mL; Pk/6
₹6,500.00INR / Pkg of 6
United Scientific Supplies Filtering Flask, Borosilicate Glass, 100 mL; Pk/6 WW-34509-46 Mfr # FG5340-100
COMPARE
100 4 1/4 Filtering Flask, Borosilicate Glass, 100 mL; Pk/6
₹10,600.00INR / Pkg of 6
United Scientific Supplies Filtering Flask, Borosilicate Glass; 250 mL WW-34509-47 Mfr # FG5340-250
COMPARE
250 6 19/64 Filtering Flask, Borosilicate Glass; 250 mL
₹2,800.00INR / Each
United Scientific Supplies Filtering Flask, Borosilicate Glass; 500 mL WW-34509-48 Mfr # FG5340-500
COMPARE
500 7 31/64 Filtering Flask, Borosilicate Glass; 500 mL
₹3,400.00INR / Each
United Scientific Supplies Filtering Flask, Borosilicate Glass; 1000 mL WW-34509-49 Mfr # FG5340-1000
COMPARE
1000 9 21/32 Filtering Flask, Borosilicate Glass; 1000 mL
₹5,100.00INR / Each
United Scientific Supplies Filtering Flask, Borosilicate Glass; 2000 mL WW-34509-50 Mfr # FG5340-2000
COMPARE
2000 12 Filtering Flask, Borosilicate Glass; 2000 mL
₹11,100.00INR / Each
United Scientific Supplies Filtering Flask, Borosilicate Glass; 5000 mL WW-34509-51 Mfr # FG5340-5000
COMPARE
5000 15 11/32 Filtering Flask, Borosilicate Glass; 5000 mL
₹34,700.00INR / Each
United Scientific Supplies Filtering Bottle, Borosilicate Glass; 10,000 mL WW-34509-52 Mfr # FFB5340-10000
COMPARE
10000 19 29/32 Filtering Bottle, Borosilicate Glass; 10,000 mL
₹63,100.00INR / Each
Showing 1- 8 of 8