United Electric NEMA 4X Pressure Switches

FEATURES
  • Legendary UE Reliability
  • Wide selection of adjustment ranges
  • Internal reference scales and adjustment lock
  • Enclosure design permits safe and easy access for wiring
  • Choice of 1/2" NPTF electrical connection or knockouts
  • Die cast aluminum enclosure, epoxy powder coated, gasketed, captive screws
88,400.00 - ₹92,900.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Min Pressure (PSI)
Availability Pricing
United Electric H100-192 Pressure Swch 15/300PSI (95317) 15/300PSI 6845699 Mfr # H100-192
COMPARE
±1.5% of adjustable range 300 15
₹92,900.00INR / Each
United Electric H100-193 Pressure Swch 20/500PSI For Nema-4 (95324) 20/500PSI 6845700 Mfr # H100-193
COMPARE
±1.5% of adjustable range 500 20
₹88,400.00INR / Each
United Electric H100-194 Pressure Switch 80/1700 (95331) 80/1700PSI 6845701 Mfr # H100-194
COMPARE
±1.5% of adjustable range 1700 80
₹88,400.00INR / Each
United Electric H100-190 Pressure Switch 5/30PSI Temperature Pressure Switch 6845702 Mfr # H100-190
COMPARE
±1.5% of adjustable range 30 5
₹88,400.00INR / Each
United Electric H100-191 NEMA 4X Pressure Switch, 10 to 100 spig 6845703 Mfr # H100-191
COMPARE
±1.5% of adjustable range 100 10
₹88,400.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat