United Electric Explosion-Proof Pressure and Vacuum Switches

FEATURES
  • Legendary UE reliability
  • Explosion-proof
  • Class I, Div 1 & 2, (Zone 1); Class II, Div 1 & 2; Class III
  • Worldwide approvals and certifications
  • Terminal block wiring
  • Dual conduits
  • Enclosure design permits safe and easy access for wiring
  • Internal adjustment

  • Die cast aluminum enclosure, epoxy powder coated and gasketed
  • 188,000.00 - ₹293,000.00INR / Each

    5 variations of this product are available.

    See All Product Options Below
    5 PRODUCT OPTIONS
    Compareitem specifications
    Max Pressure (PSI) Min Pressure (PSI) Wetted Materials
    Availability Pricing
    United Electric J120-134 Explosion-Proof Compound Switch, 30" Hg to 20 psi 6845691 Mfr # J120-134
    COMPARE
    20 14.735 316L SS
    ₹188,000.00INR / Each
    United Electric J120-183 Explosion-Proof Pressure Switch, 1 to 20 psi 6845692 Mfr # J120-183
    COMPARE
    20 1 316L SS, Viton® GLT
    ₹293,000.00INR / Each
    United Electric J120-185 Explosion-Proof Pressure Switch, 4 to 100 psi 6845694 Mfr # J120-185
    COMPARE
    100 4 316L SS, Viton® GLT
    ₹293,000.00INR / Each
    United Electric J120-186 Explosion-Proof Pressure Switch, 8 to 200 psi 6845695 Mfr # J120-186
    COMPARE
    200 8 316L SS, Viton® GLT
    ₹293,000.00INR / Each
    United Electric J120-189 Switch250/3500psi 316Lss Diaphragm Explosion-Proof Pressure Switch 6845697 Mfr # J120-189
    COMPARE
    3500 250 316L SS, Viton® GLT
    ₹194,000.00INR / Each
    Showing 1- 5 of 5

    Need Help with this product?

    Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

    Call Us At91-22-61394444
    Live chat