Tygon® 2375 Chemical-Resistant Transfer Tubing
Ultra-chemical-resistant tubing for reliable transfer of harsh fluids
  • Outstanding resistance to acids, bases, ketones, salts, and alcohols
  • Plasticizer-free, non-DEHP construction
  • Smooth inner surface improves flow and reduces particulate buildup
  • Safer disposal—does not release harmful hydrogen chloride gases
  • Ideal for chemical transfer, industrial cleaning, and sanitation applications
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Tygon® 2375 chemical transfer tubing delivers exceptional performance in environments where aggressive chemicals are present. Offering a unique combination of chemical resistance, clarity, and flexibility, this tubing is virtually unaffected by acids, bases, ketones, salts, and alcohols.
Plasticizer-free, non-DEHP construction eliminates the risk of fluid contamination while also preventing premature brittleness and cracking. A smooth inner surface enhances flow and reduces particulate buildup, while low sorption characteristics help maintain fluid purity and minimize cross-contamination.
16,400.00 - ₹113,000.00INR / Pkg of 1
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon 2375 Chemical-Resistant Tubing, 1/16" ID x 3/16" OD, 50 Ft 4852660 Mfr # AJK00003
COMPARE
1/16 3/16 1/16
₹16,600.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 3/32"  ID x 5/32" OD, 50 Ft 4852661 Mfr # AJK00004
COMPARE
3/32 5/32 1/32
₹16,400.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 1/8" ID x 1/4" OD, 50 Ft 4852662 Mfr # AJK00007
COMPARE
1/8 1/4 1/16
₹25,400.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 5/32"  ID x 7/32" OD, 50 Ft 4852663 Mfr # AJK00009
COMPARE
5/32 7/32 1/32
₹20,700.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 3/16"  ID x 5/16" OD, 50 Ft 4852664 Mfr # AJK00012
COMPARE
3/16 5/16 1/16
₹28,300.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 1/4"  ID x 3/8" OD, 50 Ft 4852665 Mfr # AJK00017
COMPARE
1/4 3/8 1/16
₹31,600.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 5/16"  ID x 7/16" OD, 50 Ft 4852666 Mfr # AJK00022
COMPARE
5/16 7/16 1/16
₹34,900.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 3/8"  ID x 1/2" OD, 50 Ft 4852667 Mfr # AJK00027
COMPARE
3/8 1/2 1/16
₹39,900.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 3/8"  ID x 5/8" OD, 50 Ft 4852668 Mfr # AJK00029
COMPARE
3/8 5/8 1/8
₹70,500.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 1/2"  ID x 3/4" OD, 50 Ft 4852669 Mfr # AJK00038
COMPARE
1/2 3/4 1/8
₹88,100.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 5/8"  ID x 7/8" OD, 50 Ft 4852670 Mfr # AJK00046
COMPARE
5/8 7/8 1/8
₹104,000.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 3/4" ID x 1" OD, 50 Ft 4852671 Mfr # AJK00053
COMPARE
3/4 1 1/8
₹113,000.00INR / Pkg of 1
Tygon 2375 Chemical-Resistant Tubing, 1" ID x 1-3/8" OD, 50 Ft 4852672 Mfr # AJK42064
COMPARE
1 1 3/8 3/16
₹97,400.00INR / Pkg of 1
Showing 1- 13 of 13