Tunair Full-Baffle Shake Flasks, 2.5L

Increase culture yields with optimum growth conditions for aerobic organisms
  • Baffle and turbo vane closure designs increase oxygenation compared to standard Erlenmeyer flasks
  • Cap with removable filter lining protects contents of flask from airborne particulates and eliminates need to flame neck
  • All components are autoclavable
  • Suitable for culturing aerobic microbes, mammalian, and plant cells
These shake flasks work with any standard orbital shaker having adjustable speeds from 150 to 400 rpm.
For the greatest increase in dissolved oxygen, the full-baffled flasks create a motion similar to that of an overhead stirrer with an axial impeller.
Cleanup is easy—just soak in a mild dish soap to remove dirt, and air dry. All pieces are autoclavable, including the filter linings. Filter linings are also replaceable, with silicone membrane and Dri-Gauze materials available.
8,900.00 - ₹83,500.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Material Material Description
Availability Pricing
Tunair SS-5013 Full-Baffle Shake Flask, 2.5L, 1 EA 0183557 Mfr # SS-5013
COMPARE
Not included Polypropylene Full-Baffle Shake Flask, 2.5L, 1 EA
₹8,900.00INR / Each
Tunair SS-8003 Full-Baffle Shake Flask, 2.5L; 4/Pk 0183559 Mfr # SS-8003
COMPARE
Not included Polypropylene Full-Baffle Shake Flask, 2.5L; 4/Pk
₹36,600.00INR / Pkg of 4
Tunair SS-6003C Full-Baffle Flask Kit (Dri-Gauze), 2.5L, 1 kit 0183561 Mfr # SS-6003C
COMPARE
Polypropylene (PP) Polypropylene Full-Baffle Flask Kit (Dri-Gauze), 2.5L, 1 kit
₹53,600.00INR / Each
Tunair SS-6003S Full-Baffle Flask Kit (Silicone), 2.5L, 1 kit 0183563 Mfr # SS-6003S
COMPARE
Polypropylene (PP) Polypropylene Full-Baffle Flask Kit (Silicone), 2.5L, 1 kit
₹83,500.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat