TSI Alnor CompuFlow Thermoanemometers
Measure inaccessible areas with 40" (102 cm) articulating probe
  • Display up to three measurements simultaneously on a large backlit LCD
  • Automatically calculate averages of all your parameters with the push of a button
These thermoanemometers measure air velocity, air volume, and temperature.
460,000.00 - ₹593,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Air Velocity Accuracy Air Velocity Resolution Max Air Velocity (ft/min)
Availability Pricing
TSI AVM430-A CompuFlow Thermoanemometer, manual datalogging 1025025 Mfr # AVM430-A
COMPARE
±3% of reading or ±3 ft/min (0.05 m/s) (whichever is greater) 1 ft/min (0.01 m/s) 6000
₹460,000.00INR / Each
TSI AVM440-A CompuFlow Thermoanemometer, auto datalogging w/humidity 1025036 Mfr # AVM440-A
COMPARE
±3% of reading or ±3 ft/min (0.05 m/s) (whichever is greater) 1 ft/min (0.01 m/s) 6000
₹593,000.00INR / Each