Troemner Class-F Metric Stainless Steel Hook on Top Test Weights
Stainless Steel Hook Weights are used in a variety of applications such as pressure, torque, and tensile strength testing
Troemner’s Stainless Steel Hook weights are manufactured from 300 series stainless steel. Each weight has the nominal value stamped into the top side of the weight.
Hook Top Only - Hook Top Weights are flat on top with a stationary hook in the center of the weight
31,900.00 - ₹204,000.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (g) Mass Size (lbs)
Availability Pricing
Troemner 12320-SW 100g Class F Stainless Steel Hook On Top Mass, NVLAP Accredited Certificate 0102369 Mfr # 12320-SW
COMPARE
NVLAP 100 0.22
₹49,100.00INR / Each
Troemner 12318-SW 200g Class F Stainless Steel Hook On Top Mass, NVLAP Accredited Certificate 0102370 Mfr # 12318-SW
COMPARE
NVLAP 200 0.44
₹49,500.00INR / Each
Troemner 12316-SW 500g Class F Stainless Steel Hook On Top Weight with NVLAP Certificate 0102371 Mfr # 12316-SW
COMPARE
NVLAP 500 1.1
₹51,300.00INR / Each
Troemner 12312-SW 1kg Class F SS Hook On Top Weight with NVLAP Accredited Certificate 0102372 Mfr # 12312-SW
COMPARE
NVLAP 1000 2.2
₹52,200.00INR / Each
Troemner 12310-SW 2kg Class F SS Hook On Top Weight with NVLAP Accredited Certificate 0102373 Mfr # 12310-SW
COMPARE
NVLAP 2000 4.4
₹85,900.00INR / Each
Troemner 12306-SW 5kg Class F S.S. Hook On Top Weight with NVLAP Accredited Certificate 0102374 Mfr # 12306-SW
COMPARE
NVLAP 5000 11
₹123,000.00INR / Each
Troemner 12304-SW 10kg Class F Stainless Steel Hook On Top Weight with NVLAP Certificate 0102375 Mfr # 12304-SW
COMPARE
NVLAP 10000 22
₹204,000.00INR / Each
Troemner 12012-SW 1 Lb Class F SS Hook On Top Weight with NVLAP Accredited Certificate 0102384 Mfr # 12012-SW
COMPARE
NVLAP 200 1
₹48,800.00INR / Each
Troemner 12006-SW 10lb Class F Stainless Steel Hook On Top Weight with NVLAP Certificate 0102386 Mfr # 12006-SW
COMPARE
NVLAP 5000 10
₹114,000.00INR / Each
Troemner 12312-S 1 kg Class F Stainless Steel Hook Top Weight with No Cert 0103409 Mfr # 12312-S
COMPARE
None 1000 2.2
₹34,000.00INR / Each
Troemner 12316-S 500 g Class F Stainless Steel Hook Top Weight with No Cert 0103411 Mfr # 12316-S
COMPARE
None 500 1.1
₹31,900.00INR / Each
Troemner 12306-ST 5 kg, Class F Stainless Steel Hook Top Weight with Traceable Certificate 0103421 Mfr # 12306-ST
COMPARE
NIST 5000 11
₹102,000.00INR / Each
Troemner 12310-ST 2 kg, Class F Stainless Steel Hook Top Weight with Traceable Certificate 0103422 Mfr # 12310-ST
COMPARE
NIST 2000 4.4
₹72,800.00INR / Each
Troemner 12312-ST 1 kg, Class F Stainless Steel Hook Top Weight with Traceable Certificate 0103423 Mfr # 12312-ST
COMPARE
NIST 1000 2.2
₹46,100.00INR / Each
Troemner 12316-ST 500 g, Class F Stainless Steel Hook Top Weight with Traceable Certificate 0103424 Mfr # 12316-ST
COMPARE
NIST 500 1.1
₹43,400.00INR / Each
Troemner 12320-ST 100 g, Class F Stainless Steel Hook Top Weight with Traceable Certificate 0103426 Mfr # 12320-ST
COMPARE
NIST 100 0.22
₹35,800.00INR / Each
Showing 1- 16 of 16