Troemner Class-6 Metric Cast Iron Slotted Weights
Use in a variety of applications such as pressure, torque, and tensile strength testing
  • Use several slotted weights together to build up from a minimum weight to a maximum test load
  • A durable coat of paint protects each weight from rusting and chipping
  • Color coded – gold for metric
Cast Iron Slotted Weights are used in a variety of applications such as pressure, torque, and tensile strength testing. Cast Iron Slotted Weights are typically used with a hanger that also has its weight calibrated so the hanger can be used as part of the overall weight under test. Several Cast Iron Slotted Weights may be used together to build up from a minimum weight to a maximum test load. The hanger weight selected should be able to accommodate the total load needed.
Cast Iron Slotted Weights above 2 kg/5 lb have adjusting cavities. Weights below 2 kg/5 lb are adjusted by removing material from the underside of the weight. Each weight has its nominal value cast into the topside of the weight.
15,200.00 - ₹52,800.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (kg) Mass Size (lbs)
Availability Pricing
Troemner 9694T 5kg, 6" Dia, Metric Slotted Weights, with Traceable Certificate 1196601 Mfr # 9694T
COMPARE
NIST/NVLAP 5 11.1
₹42,700.00INR / Each
Troemner 9692T 2kg, 5" Dia, Metric Slotted Weights, with Traceable Certificate 1196602 Mfr # 30391650
COMPARE
NIST/NVLAP 2 4.4
₹33,700.00INR / Each
Troemner 9690T 1kg 5" Dia, Metric Slotted Weights, with Traceable Certificate 1196603 Mfr # 30391649
COMPARE
NIST/NVLAP 1 2.2
₹30,200.00INR / Each
Troemner 9688T 500g, 5" Dia, Metric Slotted Weights, with Traceable Certificate 1196604 Mfr # 9688T
COMPARE
NIST/NVLAP 0.5 1.1
₹29,400.00INR / Each
Troemner 9696 10kg, 8" Dia, Metric Slotted Weights 1196691 Mfr # 9696
COMPARE
None 10 22.4
₹36,200.00INR / Each
Troemner 9694 5kg, 6" Dia, Metric Slotted Weights 1196692 Mfr # 9694
COMPARE
None 5 11.1
₹31,100.00INR / Each
Troemner 9692 2kg, 5" Dia, Metric Slotted Weights 1196693 Mfr # 9692
COMPARE
None 2 4.4
₹19,300.00INR / Each
Troemner 9690 1kg, 5" Dia, Metric Slotted Weights 1196694 Mfr # 9690
COMPARE
None 1 4.4
₹16,700.00INR / Each
Troemner 9688 500g, 5" Dia, Metric Slotted Weights 1196695 Mfr # 9688
COMPARE
None 0.5 1.1
₹16,300.00INR / Each
Troemner 9686 200g, 3-3/4" Dia, Metric Slotted Weights 1196696 Mfr # 9686
COMPARE
None 0.2 0.44
₹15,200.00INR / Each
Troemner 9696T 10kg 8" Dia Metric Slotted Weights, with Traceable Certificate 1196700 Mfr # 9696T
COMPARE
NIST/NVLAP 10 22.1
₹52,800.00INR / Each
Showing 1- 11 of 11