Troemner Class-3 Analytical Calibration Weights
ASTM Class 3 - Appropriate for calibrating balances with moderate precision and with a readability as low as 0.1 g to 0.01 g.
  • Surfaces are polished to a perfect, mirror like finish
  • Masses are less vulnerable to wear and scratches
  • Low magnetic susceptibility, uniform consistency and high resistance to corrosion
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity and turned up end for easy handling
  • 1g to 5kg weights are two-piece construction with adjusting cavity and handling knob
Appropriate for calibrating balances with moderate precision and with a readability as low as 0.1 g to 0.01 g.
The weights are manufactured of the finest stainless steels available, including Troemner Alloy 8 Stainless Steel, which is available exclusively through Troemner. This unique material possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion. Alloy 8 is the finest grade of stainless steel available and is similar to material originally specified in the manufacture of 1 kg state lab primary standards.
Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances
decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.
Suggested Markets for Analytical Precision Weights include, but are not limited to, pharmaceutical, biotech, nanotechnology, life science, forensic and nuclear industries.
23,300.00 - ₹258,000.00INR / Each
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (g)
Availability Pricing
Troemner 7012-3W 2kg Analytical Class 3 Weight with NVLAP Accredited Certificate 0102333 Mfr # 7012-3W
COMPARE
NIST/NVLAP 2000
₹258,000.00INR / Each
Troemner CLASS 3 1kg Analytical Class 3 Weight with NVLAP Certificate 0102469 Mfr # 7013-3W
COMPARE
NIST/NVLAP 1000
₹169,000.00INR / Each
Troemner CLASS 3 500g Analytical Class 3 Weight with NVLAP Certificate 0102481 Mfr # 7014-3W
COMPARE
NIST/NVLAP 500
₹89,100.00INR / Each
Troemner CLASS 3 200g Analytical Class 3 Weight with NVLAP Certificate 0102489 Mfr # 7016-3W
COMPARE
NIST/NVLAP 200
₹67,500.00INR / Each
Troemner CLASS 3 100g Analytical Class 3 Weight with NVLAP Certificate 0102508 Mfr # 7017-3W
COMPARE
NIST/NVLAP 100
₹42,500.00INR / Each
Troemner CLASS 3 50g Analytical Class 3 Weight with NVLAP Certificate 0102522 Mfr # 7018-3W
COMPARE
NIST/NVLAP 50
₹41,500.00INR / Each
Troemner CLASS 3 20g Analytical Class 3 Weight with NVLAP Certificate 0102542 Mfr # 7020-3W
COMPARE
NIST/NVLAP 20
₹39,400.00INR / Each
Troemner CLASS 3 10g Analytical Class 3 Weight with NVLAP Certificate 0102565 Mfr # 7021-3W
COMPARE
NIST/NVLAP 10
₹38,400.00INR / Each
Troemner CLASS 3 5g Analytical Class 3 Weight with NVLAP Certificate 0102573 Mfr # 7022-3W
COMPARE
NIST/NVLAP 5
₹37,300.00INR / Each
Troemner CLASS 3 2g Analytical Class 3 Weight with NVLAP Certificate 0102580 Mfr # 7024-3W
COMPARE
NIST/NVLAP 2
₹36,200.00INR / Each
Troemner CLASS 3 1g Analytical Class 3 Weight with NVLAP Certificate 0102584 Mfr # 7025-3W
COMPARE
NIST/NVLAP 1
₹34,400.00INR / Each
Troemner CLASS 3 500 mg, Analytical Class 3 Weight with NVLAP Certificate 0102589 Mfr # 7026-3W
COMPARE
NIST/NVLAP 0.5
₹30,500.00INR / Each
Troemner CLASS 3 100 mg, Analytical Class 3 Weight with NVLAP Certificate 0102599 Mfr # 7029-3W
COMPARE
NIST/NVLAP 0.1
₹30,500.00INR / Each
Troemner CLASS 3 10 mg, Analytical Class 3 Weight with NVLAP Certificate 0102616 Mfr # 7033-3W
COMPARE
NIST/NVLAP 0.01
₹30,500.00INR / Each
Troemner CLASS 3 2 mg, Analytical Class 3 Weight with NVLAP Certificate 0102644 Mfr # 7036-3W
COMPARE
NIST/NVLAP 0.002
₹30,500.00INR / Each
Troemner CLASS 3 1 mg, Analytical Class 3 Weight with NVLAP Certificate 0102656 Mfr # 7037-3W
COMPARE
NIST/NVLAP 0.001
₹30,500.00INR / Each
Troemner 7013-3T 1 kg Class 3 Analytical Weight with Traceable Cert 0103177 Mfr # 7013-3T
COMPARE
NIST 1000
₹154,000.00INR / Each
Troemner 7014-3T 500 g Class 3 Analytical Weight with Traceable Cert 0103178 Mfr # 7014-3T
COMPARE
NIST 500
₹70,500.00INR / Each
Troemner 7016-3T 200 g Class 3 Analytical Weight with Traceable Cert 0103179 Mfr # 7016-3T
COMPARE
NIST 200
₹53,000.00INR / Each
Troemner 7017-3T 100 g Class 3 Analytical Weight with Traceable Cert 0103180 Mfr # 7017-3T
COMPARE
NIST 100
₹36,900.00INR / Each
Showing 1 - 20 of 28