Troemner Class-1 Cal-Pak Calibration Mass Sets
Cal-Pak is the ideal weight set to meet the USP-41 requirements. It has a maximum and 10% weight that meet the test weight requirements and a minimum weight to test the repeatability of the balance to determine the range.
  • Low magnetic susceptibility for less interference with weighing instruments
  • Finest grade of stainless steel available—offers uniform consistency and high resistance to corrosion for long-term mass accuracy
  • Low cost of ownership—recalibration costs lower for three weights versus an entire set
Cal-Paks are designed for routine checks and verification of your balances using 3 weights to span the entire range of the balance. The weights include the manufacturers recommended calibration weight, 10% of the calibration weight, and the minimum weight. The maximum weight is used to calibrate the balance externally, the mid weight is used to test the sensitivity of the balance and the minimum weight is the lowest possible weight that can be weighed on the balance. Cal-Paks add a higher level of confidence for calibration and verification over the entire range of your balance.
Cal-Paks are supplied in a case with accessories for care and handling of the weights including gloves and forceps. The Cal-Pak comes with a flash drive containing the electronic NVLAP* Accredited Certificate. This paperless system assures you can easily access and maintain your certificates. All components of the Cal-Pak and carrying case are linked through the use of a unique serial number allowing you to trace your individual weights to the appropriate NVLAP* Accredited Certificate and carrying case. Cal-Paks are available in ANSI/ASTM E617 Class 1 with a NVLAP* Accredited Certificate
91,000.00 - ₹360,000.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Contents of Set Mass Certification Mass Class
Availability Pricing
Troemner 7255-1W 100G-200mg Cal-Pak Class 1 with NVLAP Certificate WW-11967-01 Mfr # 7255-1W
COMPARE
100G-200mg NVLAP Class 1
₹91,000.00INR / Each
Troemner 7256-1W 100G-2g Cal-Pak Class 1 with NVLAP Certificate WW-11967-02 Mfr # 7256-1W
COMPARE
100G-2g NVLAP Class 1
₹94,100.00INR / Each
Troemner 7265-1W 2kg-20g Cal-Pak Class 1 with NVLAP Certificate WW-11967-04 Mfr # 7265-1W
COMPARE
2kg-20g NVLAP Class 1
₹309,000.00INR / Each
Troemner 7253-1W 50G-2g Cal-Pak Class 1 with NVLAP Certificate WW-11967-05 Mfr # 7253-1W
COMPARE
50G-2g NVLAP Class 1
₹91,000.00INR / Each
Troemner 7257-1W 200G-200mg Cal-Pak Class 1 with NVLAP Certificate WW-11967-06 Mfr # 7257-1W
COMPARE
200G-200mg NVLAP Class 1
₹106,000.00INR / Each
Troemner 7252-1W 50G-200mg Cal-Pak Class 1 with NVLAP Certificate WW-11967-07 Mfr # 7252-1W
COMPARE
50G-200mg NVLAP Class 1
₹91,000.00INR / Each
Troemner 7264-1W 2kg-2g Cal-Pak Class 1 with NVLAP Certificate WW-11967-08 Mfr # 7264-1W
COMPARE
2kg-2g NVLAP Class 1
₹241,000.00INR / Each
Troemner 7258-1W 200G-2g Cal-Pak Class 1 with NVLAP Certificate WW-11967-09 Mfr # 7258-1W
COMPARE
200G-2g NVLAP Class 1
₹106,000.00INR / Each
Troemner 7261-1W 500G-2g Cal-Pak Class 1 with NVLAP Certificate WW-11967-10 Mfr # 7261-1W
COMPARE
500G-2g NVLAP Class 1
₹134,000.00INR / Each
Troemner 7254-1W 100G-20mg Cal-Pak Class 1 with NVLAP Certificate WW-11967-11 Mfr # 7254-1W
COMPARE
100G-20mg NVLAP Class 1
₹91,000.00INR / Each
Troemner 7262-1W 1kg-2g Cal-Pak Class 1 with NVLAP Certificate WW-11967-12 Mfr # 7262-1W
COMPARE
1kg-2g NVLAP Class 1
₹165,000.00INR / Each
Troemner 7267-1W 5kg-20g Cal-Pak Class 1 with NVLAP Certificate WW-11967-13 Mfr # 7267-1W
COMPARE
5kg-20g NVLAP Class 1
₹360,000.00INR / Each
Troemner 7260-1W 500g-200mg Cal-Pak Class 1 with NVLAP Certificate WW-11967-15 Mfr # 7260-1W
COMPARE
500g-200mg NVLAP Class 1
₹134,000.00INR / Each
Troemner 7270-1W 20g, 2g, 20 Mg Cal-Pak Class 1 with NVLAP Certificate WW-11967-34 Mfr # 7270-1W
COMPARE
20g, 2g, 20 Mg NVLAP Class 1
₹93,600.00INR / Each
Showing 1- 14 of 14