Troemner Class-0 UltraClass Analytical Calibration Weight Sets

UltraClass - The weight tolerance is equal to ANSI/ASTM E617 Class 0 and exceeds OIML R 111 Class E2. This class is used as a reference standard for calibrating other reference standards and is appropriate for calibrating highprecision analytical balances with a readability as low as 0.01 mg.
  • UltraClass Tolerance = ASTM Class 0
  • UltraClass weights, 1 g through 5 kg, are made from exclusive Troemner Alloy 8 stainless steel
These weights are developed to meet the most demanding calibration needs with the ability to be adjusted. Consistent automated measurement means that Troemner UltraClass weights are the most precise two-piece weights available with weight tolerances that equal or exceed comparable ANSI/ASTM E617 Classes.

The weights are manufactured of the finest stainless steels available, including Troemner Alloy 8 Stainless Steel, which is available exclusively through Troemner. This unique material possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion. Alloy 8 is the finest grade of stainless steel available and is similar to material originally specified in the manufacture of 1 kg state lab primary standards.


Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances
decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.

Suggested Markets for Analytical Precision Weights include, but are not limited to, pharmaceutical, biotech, nanotechnology, life science, forensic and nuclear industries.
293,000.00 - ₹805,000.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Contents of Set Mass Certification Description
Availability Pricing
Troemner 7223-0W 200g-1 mg, Ultraclass Mass Set, NVLAP Accredited Certificate 0102763 Mfr # 7223-0W
COMPARE
1 mg, two 2 mg, 5 mg, 10 mg, two 20 mg, 50 mg, 1 g, two 2 g, 5 g, 10 g, two 20 g, 50 g, 100 g, 200 g NVLAP 200g-1 mg, Ultraclass Mass Set, NVLAP Accredited Certificate
₹795,000.00INR / Each
Troemner 7219-0T 500 g-1 mg UltraClass Analytical Set with Traceable Cert 0103232 Mfr # 7219-0T
COMPARE
500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg NIST 500 g-1 mg UltraClass Analytical Set with Traceable Cert
Call For Price
Troemner 7224-0T 100 g-1 g UltraClass Analytical Set with Traceable Cert 0103237 Mfr # 7224-0T
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g NIST 100 g-1 g UltraClass Analytical Set with Traceable Cert
₹293,000.00INR / Each
Troemner 7218-0W 500 g-1 g UltraClass Analytical Set with NVLAP Cert 0103246 Mfr # 7218-0W
COMPARE
500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g NVLAP 500 g-1 g UltraClass Analytical Set with NVLAP Cert
₹805,000.00INR / Each
Troemner 7206-0T 20 kg-1 g UltraClass Analytical Set with Traceable Cert 0103495 Mfr # 7206-0T
COMPARE
20 kg, 10 kg, 5 kg, (2) 2 kg, 1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g NIST 20 kg-1 g UltraClass Analytical Set with Traceable Cert
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat