Troemner Class-0 Cal-Pak Calibration Mass Sets
Cal-Pak is the ideal weight set to meet the USP-41 requirements. It has a maximum and 10% weight that meet the test weight requirements and a minimum weight to test the repeatability of the balance to determine the range.
  • Low magnetic susceptibility for less interference with weighing instruments
  • Finest grade of stainless steel available—offers uniform consistency and high resistance to corrosion for long-term mass accuracy
  • Low cost of ownership—recalibration costs lower for three weights versus an entire set
Cal-Paks are designed for routine checks and verification of your balances using 3 weights to span the entire range of the balance. The weights include the manufacturers recommended calibration weight, 10% of the calibration weight, and the minimum weight. The maximum weight is used to calibrate the balance externally, the mid weight is used to test the sensitivity of the balance and the minimum weight is the lowest possible weight that can be weighed on the balance. Cal-Paks add a higher level of confidence for calibration and verification over the entire range of your balance.
Cal-Paks are supplied in a case with accessories for care and handling of the weights including gloves and forceps. The Cal-Pak comes with a flash drive containing the electronic NVLAP* Accredited Certificate. This paperless system assures you can easily access and maintain your certificates. All components of the Cal-Pak and carrying case are linked through the use of a unique serial number allowing you to trace your individual weights to the appropriate NVLAP* Accredited Certificate and carrying case. Cal-Paks are available in ANSI/ASTM E617 Class 1 with a NVLAP* Accredited Certificate
95,200.00 - ₹134,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (g) Description
Availability Pricing
Troemner 7250-00W 2g-1mg Cal-Pak Class 0 with NVLAP Certificate, WW-11967-14 Mfr # 7250-00W
COMPARE
NIST/NVLAP 2 2g-1mg Cal-Pak Class 0 with NVLAP Certificate,
₹95,200.00INR / Each
Troemner 7251-00W 5G-1mg Cal-Pak Class 0 with NVLAP Certificate, WW-11967-17 Mfr # 7251-00W
COMPARE
NIST/NVLAP 5 5G-1mg Cal-Pak Class 0 with NVLAP Certificate,
₹95,200.00INR / Each
Troemner 7259-1W 500G-20mg Cal-Pak Class 0 with NVLAP Certificate, WW-11967-18 Mfr # 7259-1W
COMPARE
NIST/NVLAP 500 500G-20mg Cal-Pak Class 0 with NVLAP Certificate,
₹134,000.00INR / Each