TriForest Erlenmeyer Flask Clamps
Universal replacement clamps for shaking incubators
  • Compatible with all industry-standard shaking incubators
  • Constructed of blue nonscratch polypropylene
  • Autoclavable at 121°C (249°F)
These clamps are suitable for standard shaking incubators and accept Erlenmeyer flasks. Screw the clamp on the platform and snap the flask into the clamp.
5,300.00 - ₹12,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Height (in) Description
Availability Pricing
TriForest Erlenmeyer Flask Clamp, PC, for 125 mL Shaker, each 0601937 Mfr # SFC0125
COMPARE
3 2 Erlenmeyer Flask Clamp, PC, for 125 mL Shaker
₹5,300.00INR / Each
TriForest Erlenmeyer Flask Clamp, PC, for 250 mL Shaker, each 0601938 Mfr # SFC0250
COMPARE
3.6 2.3 Erlenmeyer Flask Clamp, PC, for 250 mL Shaker
₹6,400.00INR / Each
TriForest Erlenmeyer Flask Clamp, PC, for 1000 mL Shaker, each 0601941 Mfr # SFC1000
COMPARE
5.7 3 Erlenmeyer Flask Clamp, PC, for 1000 mL Shaker
₹10,500.00INR / Each
TriForest Erlenmeyer Flask Clamp, PC, for 2000 mL Shaker, each 0601943 Mfr # SFC2000
COMPARE
6.7 3.5 Erlenmeyer Flask Clamp, PC, for 2000 mL Shaker
₹12,300.00INR / Each