Trajan Syringe Replacement Needle and Plunger Kits
Designed and tested to meet critical autosampler specifications
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
7,400.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 033715 Replacement Kit, Compatible with Agilent Plunger-in-Needle Syringes, 42 mm L, 23 G; 1/Ea 1600307 Mfr # 033715
COMPARE
₹7,400.00INR / Each
Trajan SGE 033730 Replacement Kit, Compatible with Agilent Autosamplers, 0.5 uL, 42 mm L, 23 G; 1/Ea 1600309 Mfr # 033730
COMPARE
₹7,400.00INR / Each
Trajan SGE 033745 Replacement Kit, Compatible with Shimadzu AOC Plunger-in-Needle Syringes, 23 G; 1/Ea 1600311 Mfr # 033745
COMPARE
₹7,400.00INR / Each
Trajan SGE 033765 Replacement kits, for Plunger-in-Needle Syringe, 0.5 uL, 70 mm L, 23 g, Cone Tip; 1/Ea 1600314 Mfr # 033765
COMPARE
₹7,400.00INR / Each
Trajan SGE 034715 Replacement Kit, Compatible with Agilent Autosamplers, 1 uL, 42 mm L, 23 G; 1/Ea 1600324 Mfr # 034715
COMPARE
₹7,400.00INR / Each