Trajan Replacement Parts for High Performance Valves
Offered in micro control or pneumatic: delivering gas control in the most appropriate format for your analysis
  • Minimal exposure to stainless steel
  • Fully swept flow path
  • Easy installation
  • Minimal dead volume
Designed specifically for chromatography applications. Can be used at elevated temperatures from high vacuum to medium pressure.
2,400.00 - ₹10,200.00INR / Pkg of 5
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 123681 Replacement part for use with Micro control Valve, VSV-6 Vespel Valve Seat; 5/Pk 1599721 Mfr # 123681
COMPARE
₹10,200.00INR / Pkg of 5
Trajan SGE 123687 Replacement PTFE Seals, for MOVP Valves; 2/Pk 1599722 Mfr # 123687
COMPARE
₹2,400.00INR / Pkg of 2