Trajan Removeable Cone-Tip Needle Autosampler Syringes
Designed and tested to meet critical autosampler specifications
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
7,700.00 - ₹19,200.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 001871 Removable CT Needle Syringe, Compatible with Thermo RSH, 5 uL, 57 mm L, 23 G; 1/Ea 1599917 Mfr # 001871
COMPARE
₹19,200.00INR / Each
Trajan SGE 002815 Removable Needle CT Syringe, Compatible with Agilent Autosampler, 10 uL, 23 G; 1/Ea 1599971 Mfr # 002815
COMPARE
₹9,100.00INR / Each
Trajan SGE 002871 Removable Needle BT Syringe, Compatible with Thermo RSH, 10 uL, 57 mm L, 23 G; 1/Ea 1599991 Mfr # 002871
COMPARE
₹13,900.00INR / Each
Trajan SGE 002875 Removable Needle Syringe, Compatible with CTC RTC RSH, 10 uL, 57 mm L, 26 G; 1/Pk 1599995 Mfr # 002875
COMPARE
₹14,400.00INR / Each
Trajan SGE 002897 Removable Needle Syringe, Compatible with Shimadzu Autosampler, 10 uL, 26 G; 1/Ea 1600000 Mfr # 002897
COMPARE
₹12,000.00INR / Each
Trajan SGE 002902 Removable Needle Syringe, Compatible with Shimadzu Autosampler, 10 uL, 23 G; 1/Ea 1600002 Mfr # 002902
COMPARE
₹12,400.00INR / Each
Trajan SGE 002952 Removable Needle Syringe, Compatible with Varian/Bruker, 10 uL, 50 mm L, 26 G; 1/Ea 1600006 Mfr # 002952
COMPARE
₹7,800.00INR / Each
Trajan SGE 002982 Removable Cone-Tip Needle Syringe, Compatible with TriPlus™ RSH Autosampler, 2 uL, 26 G; 1/Ea 1600018 Mfr # 002982
COMPARE
₹7,700.00INR / Each
Trajan SGE 002984 Removable Cone-Tip Needle Syringe, Compatible with TriPlus™ RSH Autosampler, 10 uL, 23 G; 1/Ea 1600020 Mfr # 002984
COMPARE
₹7,800.00INR / Each
Trajan SGE 0029841 Removable Needle Syringe, Compatible with CTC GC Autosampler, 10 uL, 23 G; 1/Ea 1600021 Mfr # 0029841
COMPARE
₹7,800.00INR / Each
Trajan SGE 002993 Removable Cone-Tip Needle Syringe, Compatible with TriPlus™ RSH Autosampler, 10 uL, 26 G; 1/Ea 1600031 Mfr # 002993
COMPARE
₹10,200.00INR / Each
Trajan SGE 0029931 Removable Cone-Tip Needle Syringe, Compatible with Thermo Scientific AS2000, 10 uL, 26 G; 1/Ea 1600032 Mfr # 0029931
COMPARE
₹10,200.00INR / Each
Trajan SGE 004665 Removable Needle Syringe, Compatible with Agilent Autosampler, 50 uL, 42mm L, 23 G; 1/Ea 1600067 Mfr # 004665
COMPARE
₹12,300.00INR / Each
Trajan SGE 031798 Removable Needle Syringe with Gas Tight plunger, Compatible with Shimadzu, 10 uL; 1/Ea 1600229 Mfr # 031798
COMPARE
₹8,100.00INR / Pkg of 1
Trajan SGE 002898 Removable Needle Shimadzu Syringe with 4.2 cm 0.63 mm OD Cone Tipped Needle, 10 uL; 1/Ea 1601134 Mfr # 002898
COMPARE
₹12,000.00INR / Each
Showing 1- 15 of 15