Trajan Removeable Cone-Tip Needle Autosampler NanoVolume Syringes
Inject down to 50nL with high precision and accuracy.
  • Plunger extends into the needle tip
  • Sample contained only in the needle, no glass contact
Ideal for dispensing very small liquid volumes. When the plunger is depressed, the sample is completely dispensed by the NanoVolume plunger that extends to the tip of the needle.
12,900.00 - ₹20,400.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 000400 Removable Needle CT Syringe, Compatible with Agilent Autosampler, 0.5 uL, 26 G; 1/Ea 1599872 Mfr # 000400
COMPARE
₹13,500.00INR / Each
Trajan SGE 000415 Removable Needle CT Syringe, Compatible with Agilent Autosampler, 0.5 uL, 23-26 G; 1/Ea 1599874 Mfr # 000415
COMPARE
₹16,800.00INR / Each
Trajan SGE 000440 Removable Needle Syringe, Compatible with Shimadzu Autosampler, 0.5uL, 42 mm L, 26 G; 1/Ea 1599875 Mfr # 000440
COMPARE
₹14,900.00INR / Each
Trajan SGE 000445 Removable Needle Syringe, Compatible with Shimadzu Autosampler,  0.5 uL, 23 G; 1/Ea 1599876 Mfr # 000445
COMPARE
₹14,900.00INR / Each
Trajan SGE 000475 Removable Needle CT Syringe, Compatible with PerkinElmer Autosampler, 0.5 uL, 26 G; 1/Ea 1599877 Mfr # 000475
COMPARE
₹17,100.00INR / Each
Trajan SGE 000478 Removable Needle CT Syringe, Compatible with PerkinElmer Autosampler, 0.5 uL, 23 G; 1/Ea 1599878 Mfr # 000478
COMPARE
₹17,100.00INR / Each
Trajan SGE 000480 Removable Needle CT Syringe, Compatible with Thermo RSH Autosampler, 0.5 uL, 23 G; 1/Ea 1599879 Mfr # 000480
COMPARE
₹20,400.00INR / Each
Trajan SGE 000490 Removable Needle CT Syringe, Compatible with Thermo TriPlus and AS3000, 0.5 uL, 26 G; 1/Ea 1599880 Mfr # 000490
COMPARE
₹14,800.00INR / Each
Trajan SGE 000492 Removable Needle CT Syringe, Compatible with Thermo TriPlus and AS3000, 0.5 uL, 23 G; 1/Ea 1599881 Mfr # 000492
COMPARE
₹14,800.00INR / Each
Trajan SGE 000610 Removable Needle Syringe, Compatible with Agilent Autosampler, 1 uL, 23 G; 1/Ea 1599890 Mfr # 000610
COMPARE
₹12,900.00INR / Each
Trajan SGE 000680 Removable CT Needle Syringe, Compatible with Thermo RSH, 1 uL, 57 mm L, 23 G; 1/Ea 1599892 Mfr # 000680
COMPARE
₹20,400.00INR / Each
Showing 1- 11 of 11