Trajan Removeable Cone-Tip Needle Autosampler Gas Tight Syringes
Designed and tested to meet critical autosampler specifications
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
9,800.00 - ₹89,800.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 002820 Gas Tight Syringe, Compatible with Agilent Autosampler, Removable CT Needle, 10 uL; 10/Pk 1599972 Mfr # 002820
COMPARE
₹89,800.00INR / Pkg of 10
Trajan SGE 002829 Gas Tight Syringe, Compatible with Agilent Autosampler, 10 uL, 23-26 G; 10/Pk 1599978 Mfr # 002829
COMPARE
₹9,800.00INR / Each
Trajan SGE 002924 Gas Tight Syringe, Compatible with Varian/Bruker, Removable Needle, 10 uL, 25 G; 1/Ea 1600003 Mfr # 002924
COMPARE
₹46,500.00INR / Each
Trajan SGE 002985 Gas Tight Syringe, Compatible with CTC/Thermo Autosampler, Removable Cone-Tip Needle, 10uL; 1/Ea 1600022 Mfr # 002985
COMPARE
₹9,900.00INR / Each
Trajan SGE 0029851 Gas Tight Syringe, Compatible with Varian/Bruker, Removable Needle, 10 uL, 26 G; 1/Ea 1600023 Mfr # 0029851
COMPARE
₹9,900.00INR / Each
Trajan SGE 005333 Gas Tight Syringe, Compatible with CTC PAL Systems, Removable Cone-Tip Needle, 100 uL, 26 G; 1/Ea 1600089 Mfr # 005333
COMPARE
₹13,000.00INR / Each