Trajan Fixed Luer Lock Manual Syringes
The comprehensive range of SGE syringes are designed and tested to meet critical specifications. We combine precision manufacturing of components and meticulous assembly, to produce syringes of the highest quality.
  • Minimal carryover
  • Gas tight
  • PTFE Plunger tip
  • Resistant to a wide range of solvents
7,200.00 - ₹25,900.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 004230 Gas Tight Syringes with Additional Replacement Needle, 50 uL, Fixed Luer Lock; 1/Ea 1600054 Mfr # 004230
COMPARE
₹7,800.00INR / Each
Trajan SGE 005229 Gas Tight Syringe, Fixed Luer Tip Needle, 100 uL; 1/Ea 1600078 Mfr # 005229
COMPARE
₹7,200.00INR / Each
Trajan SGE 005230 Gas Tight Syringe, Fixed Luer Lock Needle, 100 uL; 1/Ea 1600079 Mfr # 005230
COMPARE
₹7,700.00INR / Each
Trajan SGE 005232 Dispenser Syringe, Compatible with Hamilton MicroLab 500® Instruments, 100 uL; 1/Ea 1600080 Mfr # 005232
COMPARE
₹9,900.00INR / Each
Trajan SGE 006230 General Purpose Manual Syringe, 250 uL, Fixed Luer Lock; 1/Ea 1600111 Mfr # 006230
COMPARE
₹8,000.00INR / Each
Trajan SGE 007229 Gas Tight Syringes with Fixed Luer Tip, 500 uL; 1/Ea 1600138 Mfr # 007229
COMPARE
₹8,900.00INR / Each
Trajan SGE 007230 Gas Tight Syringes with Fixed Luer Lock, 500 uL; 1/Ea 1600139 Mfr # 007230
COMPARE
₹8,100.00INR / Each
Trajan SGE 007232 Dispenser Syringe, Compatible with Hamilton MicroLab 500® Instruments, 500 uL; 1/Ea 1600140 Mfr # 007232
COMPARE
₹9,900.00INR / Each
Trajan SGE 008020 Gas Tight Syringes with Fixed Luer Tip, 1 mL; 1/Ea 1600156 Mfr # 008020
COMPARE
₹13,500.00INR / Each
Trajan SGE 008420 Gas Tight Syringes with Fixed Luer Tip, 2.5 mL; 1/Ea 1600169 Mfr # 008420
COMPARE
₹9,200.00INR / Each
Trajan SGE 008425 Dispenser Syringe, Compatible with Hamilton MicroLab 500® Instruments, 2.5 mL; 1/Ea 1600170 Mfr # 008425
COMPARE
₹8,700.00INR / Each
Trajan SGE 009463 Gas Tight Syringe with Fixed Luer Lock, 25 mL, 1/Ea 1600209 Mfr # 009463
COMPARE
₹25,900.00INR / Each
Trajan SGE 008025 Dispenser Syringe, Compatible with Hamilton MicroLab 500® Instruments, 1 mL; 1/Ea 1601152 Mfr # 008025
COMPARE
₹9,000.00INR / Each
Trajan SGE 008762 Dispenser Syringe, Compatible with Hamilton MicroLab 500® Instruments, 5 mL; 1/Ea 1601155 Mfr # 008762
COMPARE
₹8,700.00INR / Each
Trajan SGE 008962 Dispenser Syringe, Compatible with Hamilton MicroLab 500® Instruments, 10 mL; 1/Ea 1601156 Mfr # 008962
COMPARE
₹15,500.00INR / Each
Showing 1- 15 of 15