Trajan Diamond MS Syringe Replacement Plunger Kits
Inject down to 50nL with high precision and accuracy.
  • Plunger extends into the needle tip
  • Sample contained only in the needle, no glass contact
Ideal for dispensing very small liquid volumes. When the plunger is depressed, the sample is completely dispensed by the NanoVolume plunger that extends to the tip of the needle.
7,700.00 - ₹15,900.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 000376 On-Column Syringe with Replacement Needle, 0.5 uL, 70 mm L; 1/Ea 1599870 Mfr # 000376
COMPARE
₹14,600.00INR / Each
Trajan SGE 0355180 Diamond MS Syringe Replacement Plunger, Compatible with Shimadzu Autosampler, 10 uL; 1/Ea 1600346 Mfr # 0355180
COMPARE
₹7,700.00INR / Each
Trajan SGE 0355191 Diamond MS Syringe Replacement Plunger, Compatible with CTC RTC/Thermo RSH, 25 uL; 1/Ea 1600348 Mfr # 0355191
COMPARE
₹8,600.00INR / Each
Trajan SGE 0355192 Diamond MS Syringe Replacement Plunger, Compatible with CTC RTC/Thermo RSH, 50 uL; 1/Ea 1600349 Mfr # 0355192
COMPARE
₹8,600.00INR / Each
Trajan SGE 003790 Diamond MS Syringe, Compatible with Thermo RSH/CTC RTC, Replacement Plunger, 25 uL; 1/Ea 1601144 Mfr # 003790
COMPARE
₹15,900.00INR / Each