Traceable® Wi-Fi Data Logging Refrigerator/Freezer Thermometers Compatible with TraceableLIVE® Cloud Service
Stay connected to your critical environments — wherever you go!
  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE wireless capability*
  • View current temperature conditions anywhere—cloud-based interface sends email, text, and push notifications
  • No software required—web, Android™, Apple® iOS data visibility
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
TraceableLIVE Wireless Technology allows multiple users to stay connected and monitor critical environments 24/7 wherever you go.
These data logging thermometers with Wi-Fi offer an easy and secure way to connect your smartphone, tablet, or personal computer remotely with your water baths, refrigerators, freezers, and biological/pharmaceutical lab equipment.
Once setup is complete you can securely monitor parameters 24/7, control your alarms remotely, view your data logging history, run reports in real time, and give access to other team members. No local software is required to use the cloud-based data interface. Get mobile push notifications, emails, or texts for out of range conditions, connectivity interruptions, low-battery condition or if your device is due for calibration. Large tripe-line LCD displays current and Min/Max readings.
Use the included hook-and-loop strip to mount the probe to the inside of a freezer. Micro cable on the probe permits freezer door to close on it. Mount the logger to the outside of the freezer with the included magnetic strip or use the built-in bench stand to place on countertop. A low-cost subscription is required for TraceableLIVE® use.
Plus the NIST-traceable precalibration means you get reliable measurements right out of the box, saving you time and money!
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
30,200.00 - ₹52,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Input Type Max Temperature (° C) Min Temperature (° C)
Availability Pricing
COMPARE
Two vaccine probes 60 -50
₹34,000.00INR / Each
COMPARE
Two quick disconnect bottle probes 60 -50
₹30,200.00INR / Each
COMPARE
Two bottle probes 60 -50
₹35,300.00INR / Each
COMPARE
One bottle probe and one bullet probe 60 -50
₹35,000.00INR / Each
COMPARE
Two bullet probes 60 -50
₹52,000.00INR / Each
COMPARE
Two stainless steel penetration probes 60 -50
₹37,600.00INR / Each