Traceable® Standard Remote-Probe Digital Thermometers with Calibration
Ten-ft flexible cord makes it easy to reach difficult areas
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Probe and cable may be totally immersed in liquid
  • Readings update every second
  • °F/°C switchable ranges
Magnetic back allows for placement on metal surfaces such as freezers, incubators, or water baths. Includes a flip-open stand for use on bench or desktop, and a spring fastener for clipping onto any edge.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
6,400.00 - ₹8,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
COMPARE
500 -58 Calibrated Remote Probe Thermometer; ±1.0°C accuracy (-20 to 100°C)
₹6,400.00INR / Each
COMPARE
500 -58 Calibrated Remote Probe Thermometer Ultra; ±0.5°C Accuracy at Tested Points
₹8,400.00INR / Each