Traceable® Logger-Trac™ Temperature Data Logger with Calibration
Built-in display shows measurements and alarm duration—no need to download data first
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Compact and reusable—ideal for transportation monitoring
  • Download data and prepare reports with thorough data analysis (requires software purchase)
  • IP66-rated housing for use in virtually any environment
  • Stores 7,680 temperature data points with time and date stamp
  • Only measures 2.56" x 1.34" x 0.35" and weighs 0.65 ounces
This compact, lightweight data logger is reusable and reprogrammable making it ideal for transportation monitoring. Unit comes preprogrammed to record temperature readings every 5 minutes and 33 seconds for 30 days with alarm setpoints at 2°C (low) and 8°C (high). For complete flexibility, the user-defined recording interval may be set based on the number of desired days to log (from 4 hours to 365 days) or the desired time interval (from every 2 seconds to every 24 hours). Once the memory is full the data logger will stop capturing data ensuring no data is lost. If the measurement falls outside of the user-determined alarm conditions for too long, "QUARANTINE" is shown on the display—perfect for monitoring environmental temperatures of refrigerated vaccines, chemicals, pharmaceuticals, and other perishable items.
The IP66 waterproof rating allows the units to be used in any location, even those that are damp or require washdown. Order the optional sleeves to provide other users with a quick reference of the logger settings including minimum, maximum, quarantine, logging interval and duration, start and stop date and time, and special instructions.
Software is required to download and analyze data or program individual units. One copy of the software may be used for multiple loggers and allows you to update settings including individual unit name/description, time zone settings, daylight savings time on/off, unit of measure (°F or °C), start delay, recording duration/sampling interval, quarantine/alarm settings, and user stop key enabling. Software and cradle sold separately.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Input Type Max Temperature (° C) Min Temperature (° C)
Availability Pricing
Traceable Logger-Trac™ Temperature Data Logger 1800500 Mfr # 6540TR
COMPARE
Temperature 50 -35