Traceable® See-Through Refrigerator Thermometers with Calibration
Compact design fits easily into tight locations
  • Large 1/2"H digits update every 30 seconds
  • Easily wall mount using the suction cup or hook-and-loop fastener
  • °C/°F selectable
These inexpensive refrigerator thermometers are also ideal for monitoring temperatures in storerooms, labs, offices, hoods, and cabinets. Use the suction cup on the back of the thermometer, or included hook-and-loop strips, to locate it close to materials being observed inside a refrigerator or on other surfaces.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
5,600.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy
Availability Pricing
Traceable See-Through Refrigerator Thermometer Ultra; ±0.5°C Accuracy at Tested Points 9876726 Mfr # 4158TR
COMPARE
±0.5°C at tested points; otherwise ±1.5°C
₹5,600.00INR / Each