Traceable® Printing Digital Manometers with Calibration
Print data directly from the meter
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Record up to 12,000 readings
  • Connect tubing with 1/16 to 3/16” (1.6 to 4.8 mm) inner diameter
  • Obtain fast results—0.5 second response time!
  • Freeze the display with HOLD feature
This digital manometer, vacuum/pressure gauge, can read and display 11 different units of measure. Obtain either gauge pressure/vacuum or differential pressure measurements. The backlit digital display, 5/8" H LCD, is easy to read—even from 6-ft away. Typical response time is 0.5 seconds.
Create hard copies of data—on the spot with the integrated printer. Printouts includes date and time stamp to meet strict conformity requirements. Store up to 12,000 recordings in the memory, freeze the display with the HOLD feature, and recall MIN/MAX readings.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
50,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSID)
Availability Pricing
COMPARE
5
₹50,700.00INR / Each
COMPARE
100
₹50,700.00INR / Each