Traceable® Panel-Mount Remote-Probe Digital Thermometer with Calibration
Measure temperature from up to 10-ft away!
  • Panel-mount module snaps into place for easy mounting
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Panel-mount remote probe thermometer features a 10-ft flexible cord for measuring temperatures from a distance. Versatile panel-mount module snaps into place or can be used as a stand-alone unit. Model has an easy-view, 3/4" high LCD display and records minimum and maximum readings over time.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
5,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
Traceable Panel-Mount Remote-Probe Digital Thermometer with Calibration 9020526 Mfr # 4376TR
COMPARE
0.75" Display, 3.5-digit LCD 572 -58
₹5,000.00INR / Each