Traceable® GIANT-DIGITS Radio Atomic Clocks
LCD with large digits can be viewed from 200 feet away!
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Accurate to 1/10,000 of a second
The atomic clock automatically sets correct time and date from NIST's atomic clock in Colorado. Select from PST (Pacific Standard Time), MST (Mountain Standard Time), CST (Central Standard Time), and EST (Eastern Standard Time). Simultaneously displays relative humidity along with both ambient and remote temperature for easy comparison of room temperature versus outdoor temperature. Signal reception includes four strength levels; RC synchronize process, weak signal quality, acceptable signal quality, and excellent signal quality. The weather forecast indicator includes; sunny, slightly cloudy, cloudy, and rainy along with ice alert when outdoor temperature falls between -2 and 3°C.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
16,700.00 - ₹16,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Type Certifications Description
Availability Pricing
COMPARE
Main unit: 4 AA (included); Sensor: 2 AA (not included) NIST-traceable Calibrated GIANT-DIGITS Radio Atomic Clock with Remote Sensor
₹16,800.00INR / Each
COMPARE
1 AA (included) Calibrated GIANT-DIGITS Radio Atomic Clock
₹16,700.00INR / Each