Traceable® Infrared Thermometers with Calibration
High-performance, easy-to-operate infrared gun has incredible 50:1 field of view
  • Ideal for a wide range of applications with –58 to 1832°F temperature range
  • High-perfomance IR thermometer features 50:1 field of view laser sighting
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
This IR thermometer features an enhanced accuracy of ±1.5% of reading ± 2.0°C from -20° to 200°C.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
32,700.00 - ₹42,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Emissivity Max Temperature (° F)
Availability Pricing
COMPARE
50:1 Fixed 1832
₹32,700.00INR / Each
COMPARE
50:1 Adjustable 1832
₹42,200.00INR / Each