Traceable® Flip-Stick™ Thermometers with Calibration
Multi-angle probe makes taking temperature measurements in difficult to reach places a snap
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Rugged, shockproof and drop tested to 5 feet
Flip-open design make it easy to carry around in your pocket or on the wrist strap. Meter measures 11”L when extended but folds to a nice compact 6” in length. Position stainless steel probe to any angle for easy temperature measurements. Large, ¾”H digits are easy to view at a glance. Features include switchable °F/°C, Min/Max, Hold, and low-battery indicator.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
4,500.00 - ₹7,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
COMPARE
572 -58 Calibrated Flip-Stick Thermometer Ultra; ±0.3°C Accuracy at Tested Points
₹7,700.00INR / Each
COMPARE
572 -58 Calibrated Flip-Stick Thermometer; ±1°C Accuracy
₹4,500.00INR / Each