Tissue Processing Capsules
  • Hinged design is perfectly aligned and fabricated as one piece from disposable polypropylene.
  • Open mesh construction for maximum fluid exchange
  • Durable marking surface for permanent identification
  • Available in two sizes: 8 mm D x 38 mm dia; and biopsy size, 6 mm D x 29 mm dia
  • Use embedding rings to embed capsules.
15,600.00 - ₹20,600.00INR / Pkg of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
External Depth (in) External Diameter (cm)
Availability Pricing
Bio Plas Tissue Processing Capsules, 8 mm D x 38 mm dia 4865000 Mfr # 6010
COMPARE
5/16 3.8
₹15,600.00INR / Pkg of 500
Bio Plas 6020 Tissue Processing Capsules, 6 mm D x 29 mm dia 4865010 Mfr # 6020
COMPARE
15/64 2.9
₹20,600.00INR / Pkg of 500