Tip-Out Bin Systems
Customizable to your needs
  • See contents at a glance with clear plastic construction
  • Easily access items with 45° angle opening
  • Clean by lifting bins out of housing
Access supplies quickly with bins that open to an ample 45° angle. Bins are clear and impact-resistant—locate parts and check inventory at a glance. Lift out any bin from its housing to refill or wash it with soap and water.
The single-bin is intended for stand-alone use only. Other units are uniform in width, stackable, and can be fastened to optional racks, frames, or stands. Mix and match units to suit your supply needs.
8,600.00 - ₹42,400.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Length (in) Width (in)
Availability Pricing
COMPARE
4.5 3.625 23.625
₹8,600.00INR / Each
COMPARE
6.5 5.25 23.625
₹11,300.00INR / Each
COMPARE
8.1675 6.625 23.625
₹19,300.00INR / Each
COMPARE
9.5 7.75 23.625
₹26,500.00INR / Each
COMPARE
13.875 11.75 23.625
₹42,400.00INR / Each