Thermosafe PolarPack Ice Packs
The perfect choice for shipping temperature sensitive materials such as food products, pharmaceuticals, and more!
  • An economical alternative to wet ice
Refrigerant gel packs are enclosed in 4.1 mil thick polyethylene film making them an economical alternative to wet ice. With a minimum temperature rating of 32°F (0°C), they are perfect for shipping temperature sensitive materials such as food products, pharmaceuticals, and more!
2,600.00 - ₹21,300.00INR / Case of 192
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Outside Height (in) Outside Length (in) Outside Width (in)
Availability Pricing
ThermoSafe PP3 Ice Pack, 3 oz; 192/CS 0634500 Mfr # PP3
COMPARE
5/8 3 1/4 4
₹21,300.00INR / Case of 192
ThermoSafe PP6 Ice Pack, 6 oz; 96/CS 0634501 Mfr # PP6
COMPARE
3/4 5 1/2 3 1/2
₹10,000.00INR / Case of 96
ThermoSafe PP8 Ice Pack, 8 oz; 72/CS 0634502 Mfr # PP8
COMPARE
3/4 6 1/2 3 1/2
₹8,400.00INR / Case of 72
ThermoSafe PP12 Ice Pack, 12 oz; 48/CS 0634503 Mfr # PP12CP
COMPARE
1 6 5 1/2
₹5,800.00INR / Case of 48
ThermoSafe PP16 Ice Pack, 16 oz; 36/CS 0634504 Mfr # PP16
COMPARE
1 6 1/2 5 1/2
₹5,100.00INR / Case of 36
ThermoSafe PP24 Ice Pack, 24 oz; 24/CS 0634505 Mfr # PP24
COMPARE
1 1/4 8 5 1/2
₹3,800.00INR / Case of 24
ThermoSafe PP32 Ice Pack, 32 oz; 18/CS 0634506 Mfr # PP32
COMPARE
1 1/4 10 1/2 5 1/2
₹2,600.00INR / Case of 18
ThermoSafe PP48 Ice Pack, 48 oz; 12/CS 0634507 Mfr # PP48
COMPARE
1 1/4 10 1/2 7 1/2
₹4,200.00INR / Case of 12
Showing 1- 8 of 8