Thermo Scientific Turbidity Standards for AQ4500 Turbidimeter
Ensure accurate readings with turbidity standard
  • Use with AQ4500 turbidimeter
Use standards to calibrate and validate your turbidity meter.
22,730.00 - ₹214,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Type
Availability Pricing
Thermo Scientific AC45ST Orion™ Primary Standards for AQ4500 Turbidimeter 9954552 Mfr # AC45ST
COMPARE
Primary standards: SDVB for 0, 1.0, 10, 100, 1000 NTU
₹214,000.00INR / Each
Thermo Scientific AC45FZ Formazin Standard Solution, 4000 NTU; 500 mL 9990032 Mfr # AC45FZ
COMPARE
Formazin turbidity standard
₹22,730.00INR / Each