Thermo Scientific™ S4TR TX1000 Refrigerated Benchtop Centrifuge Packages

Simplify your centrifuge operation with a convenient application-based package
  • Approved for IVD MD applications—complies with the EU IVDR (EU) 2017/746 and listed by the FDA
  • Features easy-to-use controls and excellent reliability to deliver high performance
  • Cool samples down to -20°C
  • Natural refrigerant does not contribute to ozone layer depletion
  • Set up to 6 programs
  • Accepts up to 13 different rotors
  • Quick tool-less rotor exchange
  • Select from 9 acceleration and 10 deceleration rates
  • Preset run time up to 9 hours 59 minutes
  • Data transfer via RS-232 interface
  • Space saving design helps optimize work area
  • Class ISO 6 cleanroom compatible
Each S4TR TX1000 benchtop centrifuge package comes with the necessary components to run a specific protocol in one convenient bundle. Centrifuge combines intuitive controls, excellent reliability, and ease of use to maximize productivity, making it well suited for biopharmaceutical, academic research, and clinical diagnostic applications. Simplified operation allows up to six programmable methods, temperature adjustment in 1° increments, and speed control in 50 rpm increments or g force in 50 × g increments. A clear LCD display provides detailed operating parameters and real time process status, further enhancing efficiency and productivity.

Use the refrigerated centrifuge to cool samples down to –20°C—ideal for temperature-sensitive samples. The centrifuge features a natural gas hydrocarbon refrigerant (R290) that does not deplete the ozone layer and breaks down naturally reducing the environmental impact. Run centrifuge continuously, in pulse mode, or use the countdown timer to operate unattended. Select from 13 rotor options to suit your application. Auto-Lock™ rotor exchange feature provides a fast and secure push-button rotor exchange with no need for tools.

Compact ergonomic design takes up minimal benchtop space and offers easy sample loading and unloading. Safety features include SMARTSpin imbalance detection and finger-pinch prevention.
5,114,000.00INR / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Application
Availability Pricing
Thermo Scientific S4TR TX1000 Refrigerated Benchtop Centrifuge Cell Culture Package, IVD-MD; 120V-U 3616065 Mfr # 75015824
COMPARE
Cell culture
₹5,114,000.00INR / Each
Thermo Scientific S4TR TX1000 Refrigerated Benchtop Centrifuge Blood Processing Package, IVD-MD; 120V-U 3616066 Mfr # 75015825
COMPARE
Blood processing
₹5,114,000.00INR / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat