Thermo Scientific RT2 Advanced Hot Plate Stirrer
Accurate PT100 sensor provides real-time temperature detection for sensitive experimental procedures
  • Ceramic-coated aluminum top provides excellent temperature uniformity and is corrosion resistant
  • Overheat prevention circuit turns off the heater if the top plate temperature reaches 842°F (450°C)
  • "Hot top" warning visually alerts lab personnel of potential hazard
  • Digital control with PID microprocessor for easy operation
  • Three user-selectable temperature control modes: optimal, fast, and slow
  • Two-user selective timer; showing actual and set value simultaneously
  • User-controlled temperature limit setting
  • Low-profile design saves valuable bench space
  • Optional nonslip heating bath and transparent safety shield (sold separately)
  • Optional supporting rod and clamps (sold separately) allow flexibility in configuring other test equipment and accessories
360,000.00 - ₹362,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Thermo Scientific 88880006 RT2 Advanced Stirring Hot Plate, Ceramic Top; 120V 8657606 Mfr # 88880006
COMPARE
120
₹360,000.00INR / Each
Thermo Scientific 88880005 RT2 Advanced Stirring Hot Plate, Ceramic Top; 230V 8657607 Mfr # 88880005
COMPARE
230
₹362,000.00INR / Each