Thermo Scientific RT Touch Stirrers
Low-profile, lightweight design with small footprint
  • Programmable digital control
  • Timer for rundown from 99 hours 59 minutes, or operate in continuous mode
These digital stirrers provide a strong magnetic coupling that ensures consistent stirring and a gradual start-up to prevent splashing. Durable, chemical-resistant polypropylene top and body are easy to clean. The brushless DC motor will not generate heat that might harm your samples. Digital speed control is adjustable from 30 to 2000 rpm. Available in two sizes to fit your labs needs.
149,000.00 - ₹169,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Stirring Volume (Liters) Power (VAC)
Availability Pricing
Thermo Scientific 88880013 RT Touch Stirrer, Medium, 100-240 VAC; US Plug. 8657608 Mfr # 88880013
COMPARE
4 120 / 240
₹149,000.00INR / Each
Thermo Scientific 88880015 RT Touch Stirrer, Large, 100-240 VAC; US Plug 8657611 Mfr # 88880015
COMPARE
5 120 / 240
₹165,000.00INR / Each
Thermo Scientific 88880016 RT Touch Stirrer, Large, 100-240 VAC; International Plug. 8657612 Mfr # 88880016
COMPARE
5 120 / 240
₹169,000.00INR / Each