Thermo Scientific Orion Star A200 Series pH/mV/RmV/Temperature Meter
Durability and depenability make it the ideal choice for every sample, every day, anywhere
  • Multilanguage interface supports Engliah, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 1000 data points with time date stamp
  • Individual method passwords provide better security
  • Four AA batteries provide over 800 hours of field use, or order optional AC adapter for lab use
Thermo Scientific Orion Star A221 pH, mV, RmV and temperature meter features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD, shows measurement and temperature along with electrode status, time/date, sample ID, user ID, and calibration data.
Up to five point automatic calibration with USA/NIST and DIN buffers, auto-buffer recognition buffers, and recailbration feature to fix calibration errors. GLP-related features include 2000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
111,530.00 - ₹221,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max mV (mV) Max pH (pH) Description
Availability Pricing
Thermo Scientific STAR A221 Star A221 pH Portable Meter Only 5882516 Mfr # STARA2210
COMPARE
1999 20 Star A221 pH Portable Meter Only
₹111,530.00INR / Each
Thermo Scientific STAR A221 Star A221 pH Portable Meter Kit 5882518 Mfr # STARA2215
COMPARE
1999 20 Star A221 pH Portable Meter Kit
₹221,400.00INR / Each