Thermo Scientific Orion Star A 300 RDO/DO and pH/RDO/DO Meters
Advanced features and dependability makes it the top choice for measuring samples
  • Multilanguage interface supports Engliah, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 5000 data points with time date stamp
  • Individual method passwords provide better security
  • Four AA batteries provide over 800 hours of field use, or order optional AC adapter for lab use
Thermo Scientific Orion Star A 323 RDO/DO and A326 pH/RDO/DO meters features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD, shows measurement and temperature along with electrode status, time/date, sample ID, user ID, and calibration data.
GLP-related features include 5000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
Available as meter only and as complete meter kit configurations
188,890.00 - ₹425,890.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific STAR A323 Star A323 RDO/DO Portable Meter Only 5882560 Mfr # STARA3230
COMPARE
Star A323 RDO/DO Portable Meter Only
₹188,890.00INR / Each
Thermo Scientific STAR A323 Star A323 RDO/DO Portable Meter Kit 5882562 Mfr # STARA3235
COMPARE
Star A323 RDO/DO Portable Meter Kit
₹337,090.00INR / Each
Thermo Scientific STAR A326 Star A326 pH/DO Portable Meter Kit 5882574 Mfr # STARA3265
COMPARE
Star A326 pH/DO Portable Meter Kit
₹425,890.00INR / Each