Thermo Scientific™ Orion™ Conductivity Calibration Solutions
Smaller bottles are easier to carry
  • Minimize wasted solution
  • Provide more volume than pouches
Orion conductivity calibration solutions are available in portable 5 x 60 mL bottles. Portable calibration solutions are easier to carry than larger bottles, minimize wasted solution, and provide more volume than pouches. These standard solutions work with any manufacturer's conductivity meters.
22,700.00 - ₹53,800.00INR / Pkg of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Conductivity Value (µS) Description
Availability Pricing
Thermo Scientific 011008 Conductivity Standard 100 uS, 5 X 60 ml 1970490 Mfr # 011008
COMPARE
100 Conductivity Standard 100 uS, 5 X 60 ml
₹53,800.00INR / Pkg of 1
Thermo Scientific 011007 Conductivity Standard 1413 uS, 5 X 60 ml 1970491 Mfr # 011007
COMPARE
1413 Conductivity Standard 1413 uS, 5 X 60 ml
₹24,600.00INR / Pkg of 1
Thermo Scientific 011006 Conductivity Standard 12.9 mS, 5 X 60 ml 1970492 Mfr # 011006
COMPARE
12900 Conductivity Standard 12.9 mS, 5 X 60 ml
₹22,700.00INR / Pkg of 1
Thermo Scientific 011005 Conductivity Standard 111.9 mS, 5 X 60 ml 1970493 Mfr # 011005
COMPARE
111900 Conductivity Standard 111.9 mS, 5 X 60 ml
₹37,900.00INR / Pkg of 1
Thermo Scientific 990106 Conductivity Standard 12.9 mS, 475 mL 1970494 Mfr # 990106
COMPARE
12900 Conductivity Standard 12.9 mS, 475 mL
₹50,800.00INR / Each