Thermo Scientific™ Nunc™ Microplate Covers
Secure samples within your 96-well microplates
  • Protect samples from contamination and evaporation during assay processing, incubation or storage
Choose from a range of options to prevent contamination and evaporation of well contents and safely store samples within your 96-well plates.
Lids have notices for quick orientation and have individual condensation rings for enhanced sample security.
Sealing tap is an adhesive seal that effectively seals all microplate formats. Pressure sensitive adhesive is not tacky to touch but forms a strong bond to the plate upon application of pressure.
Cap Mats create a tight seal but are easy to peel off without aerosols—reducing the risk of well-to-well contamination. Caps are pierceable by hypodermic needles without compromising seal.
65,500.00 - ₹149,000.00INR / Case of 50
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Plate Type Well Shape
Availability Pricing
Thermo Scientific Nunc 264122 Microplate Lid, 96-Well, Polystyrene, Sterile; 50/Cs 0192911 Mfr # 264122
COMPARE
Polystyrene Lid All Well Shapes
₹65,500.00INR / Case of 50
Thermo Scientific Nunc 236366 Microplate Sealing Tape, Polyester, Sterile, 80 x 134 mm; 200/Cs 0192912 Mfr # 236366
COMPARE
Polyester Sealing Tape All Well Shapes
₹91,200.00INR / Case of 200
Thermo Scientific Nunc 276000 Microplate Cap Mat, 96-Well, TPE, Sterile; 50/Cs 0192954 Mfr # 276000
COMPARE
Thermoplastic elastomers (TPE) Cap Mat Round
₹149,000.00INR / Case of 50