Thermo Scientific Nalgene Straight-Side, Wide-Mouth Jars, PC
Choose clear polycarbonate or for better chemical resistance, translucent polypropylene. All containers include polypropylene screw closures. Closures are grooved for easy stacking. Autoclavable (autoclave pieces separately).
6,600.00 - ₹23,300.00INR / Pkg of 4
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene 2116-0060 Straight-sided Polycarbonate Wide-Mouth Jar, 60 mL 0676052 Mfr # 2116-0060
COMPARE
60 53 mm Polycarbonate(PC)
₹6,600.00INR / Pkg of 4
Thermo Scientific Nalgene 2116-0125 Straight-sided Polycarbonate Wide-Mouth Jar, 125 mL 0676054 Mfr # 2116-0125
COMPARE
125 70 mm Polycarbonate(PC)
₹12,300.00INR / Pkg of 4
Thermo Scientific Nalgene 2116-0250 Straight-sided Polycarbonate Wide-Mouth Jar, 250 mL 0676056 Mfr # 2116-0250
COMPARE
250 70 mm Polycarbonate(PC)
₹14,100.00INR / Pkg of 4
Thermo Scientific Nalgene 2116-0500 Straight-sided Polycarbonate Wide-Mouth Jar, 500 mL 0676058 Mfr # 2116-0500
COMPARE
500 120 mm Polycarbonate(PC)
₹22,300.00INR / Pkg of 4
Thermo Scientific Nalgene 2116-1000 Straight-sided Polycarbonate Wide-Mouth Jar, 1000 mL 0676060 Mfr # 2116-1000
COMPARE
1000 120 mm Polycarbonate(PC)
₹23,300.00INR / Pkg of 4