Thermo Scientific Nalgene Sterile Filter Unit Receivers
Use these radiation-sterilized receivers to store filtrate
  • Noncytotoxic
  • Nonpyrogenic
Receivers feature tapered sides and "grip dimples" for easy handling and a 1 1/2-turn threaded screw closure that provides a leakproof seal. Bottles have 45-mm neck.
32,200.00 - ₹50,600.00INR / Pkg of 24
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
Thermo Scientific Nalgene 455-0150 Filter Unit Receiver; 150 mL 0673140 Mfr # 455-0150
COMPARE
150
₹43,600.00INR / Pkg of 24
Thermo Scientific Nalgene 455-0250 Filter Unit Receiver; 250 mL, 24/pk 0673141 Mfr # 455-0250
COMPARE
250
₹50,600.00INR / Case of 24
Thermo Scientific Nalgene 455-0500 Filter Unit Receiver; 500 mL 0673142 Mfr # 455-0500
COMPARE
500
₹32,200.00INR / Pkg of 12
Thermo Scientific Nalgene 455-1000 Filter Unit Receiver; 1000 mL 0673144 Mfr # 455-1000
COMPARE
1000
₹49,200.00INR / Pkg of 12