Thermo Scientific Nalgene Quick Filling/Venting Closures, PP

Ideal for filling/dispensing into and out of carboys and bottles with an 83B neck finish. Quick disconnect tubing ports make installation a snap. Order replacement straight hose barb inserts and optional elbow hose barb inserts below. Note: Polyproplene is autoclavable.
69,600.00 - ₹97,500.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Material Tubing ID
Availability Pricing
Thermo Scientific Nalgene 2158-0021 Quick Filling/Venting Closures, PP, 83b 2-port 1/4" 6250800 Mfr # 2158-0021
COMPARE
Polypropylene(PP) 6.4 mm
₹69,600.00INR / Each
Thermo Scientific Nalgene 2158-0022 Quick Filling/Venting Closures, PP, 83b 2-port 3/8" 6250802 Mfr # 2158-0022
COMPARE
Polypropylene(PP) 9.5 mm
₹69,600.00INR / Each
Thermo Scientific Nalgene 2158-0031 Quick Filling/Venting Closures, PP, 83b 3-port 1/4" 6250810 Mfr # 2158-0031
COMPARE
Polypropylene(PP) 6.4 mm
₹97,500.00INR / Each
Thermo Scientific Nalgene 2158-0032 Quick Filling/Venting Closures, PP, 83b 3-port 3/8" 6250812 Mfr # 2158-0032
COMPARE
Polypropylene(PP) 9.5 mm
₹95,100.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat