Thermo Scientific Nalgene Quick-Action Spigots
Use these spigots with NALGENE carboys. Spigots have 1-1/8"-12 UNF threads and accept 5/8" tubing. Spigot 06073-22 is made of polypropylene and includes an adapter for 1/4" or 1/2" ID tubing. Spigot 06073-23 is made of TEFZEL ETFE; does not include a tubing adapter. NALGENE.
11,000.00 - ₹28,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Autoclavable Material Description
Availability Pricing
Thermo Scientific Nalgene 6422-0010 Replacement, polypropylene spigot WW-06070-22 Mfr # 6422-0010
COMPARE
Yes Polypropylene(PP) Replacement, polypropylene spigot
₹11,000.00INR / Each
Thermo Scientific Nalgene 6432-0010 quick-action Tefzel ETFE spigot WW-06070-23 Mfr # 6432-0010
COMPARE
Yes Ethylene Tetrafluoroethylene (ETFE) quick-action Tefzel ETFE spigot
₹28,100.00INR / Each