Thermo Scientific™ Nalgene™ Pipette Cleaning Baskets
Protect glass pipets from breakage while cleaning
  • Constructed of durable HDPE
  • For use with Thermo Scientific™ Nalgene™ pipette jars and washers
Protect glass pipets from breakage by using resilient, chemical-resistant Thermo Scientific™ Nalgene™ HDPE Pipet Baskets. A perforated screen in the bottom allows cleaning solutions to fill the chamber from the bottom and is recessed 0.75″ (1.9cm) for complete drainage. Use these high-denisty polyethylene baskets inside Nalgene pipet jars to soak pipets in cleaning or disinfection solutions, then drain and transfer into the Nalgene pipet washer for automatic pipet washing and rinsing.
79,000.00 - ₹102,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Compatible with Wetted Matterial Description
Availability Pricing
Thermo Scientific Nalgene DS5241-0040 Pipette Cleaning Basket for 16" L Pipets 0615000 Mfr # DS5241-0040
COMPARE
Pipette jar 06172-00 and pipette washer 06185-00 HDPE Pipette Cleaning Basket for 16" L Pipets
₹79,000.00INR / Each
Thermo Scientific Nalgene DS5241-0050 Pipette Cleaning Basket for 24" L Pipets 0615500 Mfr # DS5241-0050
COMPARE
Pipette jar 06176-00 and pipette washer 06190-00 HDPE Pipette Cleaning Basket for 24" L Pipets
₹102,000.00INR / Each