Thermo Scientific Nalgene Cellulose Nitrate (CN) Filter Units
Provide fast flow rates for buffers, salts, and other aqueous solutions
  • Triton-free CN membranes have excellent wetting properties — ideal for filtering and clarifying solutions when protein binding is not a concern
Nalgene® Filter Units provide a complete vacuum filtration system designed for the filtration of cell culture media, biological fluids, and other aqueous solutions. Each filter unit consists of polystyrene housing with a welded filter membrane mounted to a support plate in the upper structure. Slide arms with quick-disconnect tubing adapters simplify vacuum line attachment. The upper structures are easily separated from their receivers and a sterile, leakproof closure is included for filtrate storage.
46,900.00 - ₹133,000.00INR / Pkg of 12
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Pore Size (µm)
Availability Pricing
Thermo Scientific Nalgene 121-0020 Filter Units, 0.2um CN, 115 mL; 12/Pk 0673022 Mfr # 121-0020
COMPARE
115 0.2
₹46,900.00INR / Pkg of 12
Thermo Scientific Nalgene 121-0045 Filter Units, Gridded 0.45um CN, 115 mL; 12/Pk 0673040 Mfr # 121-0045
COMPARE
115 0.45
₹46,900.00INR / Pkg of 12
Thermo Scientific Nalgene 125-0045 Filter Units, 0.45um CN, 150 mL; 12/Pk 0673045 Mfr # 125-0045
COMPARE
150 0.45
₹54,400.00INR / Pkg of 12
Thermo Scientific Nalgene 125-0020 Filter Units, 0.20um CN, 150 mL; 12/Pk 0673046 Mfr # 125-0020
COMPARE
150 0.2
₹53,400.00INR / Pkg of 12
Thermo Scientific Nalgene 450-0020 Filter Units, 0.2um CN, 500 mL; 12/Pk 0673080 Mfr # 450-0020
COMPARE
500 0.2
₹102,000.00INR / Pkg of 12
Thermo Scientific Nalgene 450-0045 Filter Units, Gridded 0.45um CN, 500 mL; 12/Pk 0673090 Mfr # 450-0045
COMPARE
500 0.45
₹98,900.00INR / Pkg of 12
Thermo Scientific Nalgene 127-0020 Filter Units, Gridded 0.2um CN, 1000 mL; 12/Pk 0673092 Mfr # 127-0020
COMPARE
1000 0.2
₹131,000.00INR / Pkg of 12
Thermo Scientific Nalgene 127-0045 Filter Units, Gridded 0.45um CN, 1000 mL; 12/Pk 0673094 Mfr # 127-0045
COMPARE
1000 0.45
₹133,000.00INR / Pkg of 12
Showing 1- 8 of 8