Thermo Scientific™ Nalgene™ 489 Linear LDPE Tubing
Economically priced semirigid tubing is great for laboratory and industrial applications
  • Nontoxic, odorless, and tasteless
  • Complies with FDA regulations for food use
  • Translucent tubing makes it easy to view fluid flow
  • 50 Shore D durometer
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
The 489 linear LDPE tubing exhibits excellent chemical resistance, making it ideal for various laboratory and industrial applications. This nontoxic, odorless, and tasteless tubing complies with 21 CFR 177.1520 for food packaging.
17,100.00 - ₹54,900.00INR / Pkg of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Thermo Scientific Nalgene 8010-0125 489 Linear LDPE Tubing, 1/8" ID x 1/4" OD; 100 Ft 5014177 Mfr # 8010-0125
COMPARE
1/8 1/4 1/16
₹17,100.00INR / Pkg of 1
Thermo Scientific Nalgene 8010-0250 489 Linear LDPE Tubing, 1/4" ID x 3/8" OD; 100 Ft 5014178 Mfr # 8010-0250
COMPARE
1/4 3/8 1/16
₹24,200.00INR / Pkg of 1
Thermo Scientific Nalgene 8010-0375 489 Linear LDPE Tubing, 3/8" ID x 1/2" OD; 100 Ft 5014179 Mfr # 8010-0375
COMPARE
3/8 1/2 1/16
₹32,800.00INR / Pkg of 1
Thermo Scientific Nalgene 8010-0500 489 Linear LDPE Tubing, 1/2" ID x 5/8" OD; 100 Ft 5014180 Mfr # 8010-0500
COMPARE
1/2 5/8 1/16
₹44,200.00INR / Pkg of 1
Thermo Scientific Nalgene 8010-0625 489 Linear LDPE Tubing, 5/8" ID x 3/4" OD; 100 Ft 5014181 Mfr # 8010-0625
COMPARE
5/8 3/4 1/16
₹54,900.00INR / Pkg of 1