Thermo Scientific™ Hypersil™ CPS Classical Cyano HPLC Columns
High-efficiency columns with proven results and reproducibility
  • Suitable for Cyano phase
Thermo Scientific™ Hypersil™ columns have been trusted by researchers for over 30 years. Each batch of silica produced for these columns is tested to make sure it results in the same separations as previous batches, guaranteeing you have the same results time after time, even after changing your column. The wide range of phases ensures a column is available to match your application.
Hypersil CPS and CPS-2 (Cyano) Columns provide a more rapid mobile phase equilibration compared to silica. These columns are one of the least retentive reversed-phase materials and are useful in the separation of polar compounds. Hypersil CPS-2 is endcapped. Stable in pH range of 2 to 8.
261,000.00 - ₹359,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil 30803-154630 CPS HPLC Column, Cyano, 3 µm, 150 mm L x 4.6 mm ID 0188924 Mfr # 30803-154630
COMPARE
4.6 150 3
₹359,000.00INR / Each
Thermo Scientific 30805-254630 CPS HPLC Column, Cyano, 5 µm, 250 mm L x 4.6 mm ID 0188943 Mfr # 30805-254630
COMPARE
4.6 250 5
₹261,000.00INR / Each